siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135562.1 | NP_001129034.1 | 6 | 502 | ||
| NM_021262.2 | NP_067085.1 | 6 | 469 | |||
| XM_006239965.2 | XP_006240027.1 | 6 | 498 | |||
| XM_008764572.1 | XP_008762794.1 | 6 | 502 | |||
| GenBank mRNA | AF171072.1 | - | 6 | 432 | ||
| BC062229.1 | - | 6 | 469 | 6918194 | ||
| BC158763.1 | - | 6 | 502 | 7109165 | ||
| BC161990.1 | - | 6 | 448 | 9032684 | ||
| FQ213063.1 | - | 6 | 449 | |||
| FQ214806.1 | - | 5 | 337 | |||
| FQ215147.1 | - | 6 | 440 | |||

Acp1
Genomic Map