siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135562.1 | NP_001129034.1 | 4, 5 | 368 | ||
| NM_021262.2 | NP_067085.1 | 4, 5 | 335 | |||
| XM_006239965.2 | XP_006240027.1 | 4, 5 | 364 | |||
| XM_008764572.1 | XP_008762794.1 | 4, 5 | 368 | |||
| GenBank mRNA | AF171072.1 | - | 4, 5 | 298 | ||
| BC062229.1 | - | 4, 5 | 335 | 6918194 | ||
| BC158763.1 | - | 4, 5 | 368 | 7109165 | ||
| BC161990.1 | - | 4, 5 | 314 | 9032684 | ||
| FQ213063.1 | - | 4, 5 | 315 | |||
| FQ214806.1 | - | 3, 4 | 203 | |||
| FQ215147.1 | - | 4, 5 | 306 | |||

Acp1
Genomic Map