siRNA Details
| Gene Symbol: Acr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012490.2 | NP_036622.2 | 5 | 1061 | ||
| XM_008765665.1 | XP_008763887.1 | 6 | 2258 | |||
| XM_008765666.1 | XP_008763888.1 | 6 | 1783 | |||
| GenBank mRNA | BC097345.1 | - | 5 | 1061 | 7455117 | |
| X59254.1 | - | 5 | 1062 | |||

Acr
Genomic Map