siRNA Details
Gene Symbol: Adra1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016991.2 | NP_058687.2 | 1 | 636 | ||
GenBank mRNA | BC070920.1 | - | 1 | 636 | 7105608 | |
M60655.1 | - | 1 | 261 | |||
X51585.1 | - | 1 | 487 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
