siRNA Details
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012493.2 | NP_036625.2 | 10 | 1300 | ||
| GenBank mRNA | BC097344.1 | - | 10 | 1283 | 7459964 | |
| DY310963.1 | - | 2 | 171 | |||
| DY317147.1 | - | 5 | 631 | |||
| FQ210921.1 | - | 3 | 328 | |||
| V01254.1 | - | 7 | 1013 | |||
| X02361.1 | - | 10 | 1304 | |||

Afp
Genomic Map