siRNA Details
| Gene Symbol: Ak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033967.2 | NP_001029139.1 | 4 | 471 | ||
| NM_030986.2 | NP_112248.2 | 4 | 471 | |||
| GenBank mRNA | BC061727.1 | - | 4 | 425 | 5598607 | |
| CO570724.1 | - | 4 | 471 | |||
| D13061.1 | - | 4 | 438 | |||
| FQ228915.1 | - | 2 | 132 | |||

Ak2
Genomic Map