siRNA Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012497.1 | NP_036629.1 | 5 | 530 | ||
| GenBank mRNA | BC099749.1 | - | 5 | 516 | 5599032 | |
| FQ213506.1 | - | 5 | 533 | |||
| FQ214420.1 | - | 5 | 556 | |||
| FQ228893.1 | - | 5 | 530 | |||
| X06984.1 | - | 5 | 530 | |||

Aldoc
Genomic Map