siRNA Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012500.1 | NP_036632.1 | 14 | 1246 | ||
| XM_008766476.1 | XP_008764698.1 | 13 | 1350 | |||
| GenBank mRNA | BC166605.1 | - | 14 | 1252 | 7937766 | |
| J04733.1 | - | 14 | 1246 | |||

Apeh
Genomic Map