siRNA Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012500.1 | NP_036632.1 | 19 | 1842 | ||
| XM_008766476.1 | XP_008764698.1 | 18 | 1946 | |||
| GenBank mRNA | BC166605.1 | - | 19 | 1848 | 7937766 | |
| J04733.1 | - | 19 | 1842 | |||

Apeh
Genomic Map