siRNA Details
| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012502.1 | NP_036634.1 | 1 | 2030 | ||
| GenBank mRNA | M20133.1 | - | 1 | 2030 | ||
| M23264.1 | - | 1 | 1069 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ar| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012502.1 | NP_036634.1 | 1 | 2030 | ||
| GenBank mRNA | M20133.1 | - | 1 | 2030 | ||
| M23264.1 | - | 1 | 1069 | |||