siRNA Details
| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012504.1 | NP_036636.1 | 18 | 2767 | ||
| GenBank mRNA | BC061968.1 | - | 18 | 2778 | 5598014 | |
| FQ213537.1 | - | 3 | 370 | |||
| FQ220647.1 | - | 2 | 217 | |||
| FQ229594.1 | - | 2 | 189 | |||
| M14511.1 | - | 18 | 2767 | |||
| M28647.1 | - | 18 | 2790 | |||
| X05882.1 | - | 18 | 2544 | |||

Atp1a1
Genomic Map