siRNA Details
| Gene Symbol: Atp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012510.2 | NP_036642.2 | 4 | 716 | ||
| XM_008771385.1 | XP_008769607.1 | 4 | 541 | |||
| GenBank mRNA | M35535.1 | - | 4 | 716 | ||
| M55655.1 | - | 4 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp4b| Gene Symbol: Atp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012510.2 | NP_036642.2 | 4 | 716 | ||
| XM_008771385.1 | XP_008769607.1 | 4 | 541 | |||
| GenBank mRNA | M35535.1 | - | 4 | 716 | ||
| M55655.1 | - | 4 | 552 | |||