siRNA Details
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270630.1 | NP_001257559.1 | 2 | 1033 | ||
NM_001270631.1 | NP_001257560.1 | 2 | 1184 | |||
NM_001270632.1 | NP_001257561.1 | 2 | 1101 | |||
NM_001270633.1 | NP_001257562.1 | 2 | 1025 | |||
NM_001270634.1 | NP_001257563.1 | 3 | 912 | |||
NM_001270635.1 | NP_001257564.1 | 2 | 911 | |||
NM_001270636.1 | NP_001257565.1 | 2 | 897 | |||
NM_001270637.1 | NP_001257566.1 | 2 | 889 | |||
NM_001270638.1 | NP_001257567.1 | 2 | 766 | |||
NM_012513.4 | NP_036645.2 | 2 | 1324 | |||
XM_006234684.2 | XP_006234746.1 | 2 | 1032 | |||
XM_008762078.1 | XP_008760300.1 | 2 | 2958 | |||
GenBank mRNA | AY559248.1 | - | 2 | 827 | ||
AY559249.1 | - | 2 | 841 | |||
AY559250.1 | - | 3 | 913 | |||
BC087634.1 | - | 2 | 1019 | 7319966 | ||
EF125675.1 | - | 2 | 1324 | |||
EF125676.1 | - | 2 | 890 | |||
EF125677.1 | - | 2 | 1102 | |||
EF125678.1 | - | 2 | 1185 | |||
EF125679.1 | - | 2 | 1025 | |||
EF125680.1 | - | 2 | 1033 | |||
EF125686.1 | - | 2 | 911 | |||
EF125687.1 | - | 2 | 766 | |||
EF125688.1 | - | 2 | 898 | |||
EF125689.1 | - | 2 | 962 | |||
EF125690.1 | - | 1 | 1156 | |||
KC855561.1 | - | 1 | 194 | |||
M61175.1 | - | 2 | 736 | |||
X67108.1 | - | 1 | 685 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
