siRNA Details
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012515.2 | NP_036647.1 | 3 | 287 | ||
GenBank mRNA | FQ228784.1 | - | 3 | 289 | ||
J05122.1 | - | 2 | 230 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012515.2 | NP_036647.1 | 3 | 287 | ||
GenBank mRNA | FQ228784.1 | - | 3 | 289 | ||
J05122.1 | - | 2 | 230 |