siRNA Details
| Gene Symbol: Tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012515.2 | NP_036647.1 | 4 | 500 | ||
| GenBank mRNA | FQ228784.1 | - | 4 | 502 | ||
| J05122.1 | - | 3 | 443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspo| Gene Symbol: Tspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012515.2 | NP_036647.1 | 4 | 500 | ||
| GenBank mRNA | FQ228784.1 | - | 4 | 502 | ||
| J05122.1 | - | 3 | 443 | |||