siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 15, 16 | 2386 | ||
| XM_008760747.1 | XP_008758969.1 | 16, 17 | 2571 | |||
| XM_008760749.1 | XP_008758971.1 | 16, 17 | 2556 | |||
| XM_008760750.1 | XP_008758972.1 | 16, 17 | 2574 | |||
| GenBank mRNA | AY230250.1 | - | 15, 16 | 2388 | ||

C6
Genomic Map