siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 9 | 1316 | ||
| GenBank mRNA | AY230250.1 | - | 9 | 1318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C6| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176074.3 | NP_788263.1 | 9 | 1316 | ||
| GenBank mRNA | AY230250.1 | - | 9 | 1318 | ||