siRNA Details
Gene Symbol: Camk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042354.1 | NP_001035813.1 | 20 | 1658 | ||
NM_001042356.1 | NP_001035815.1 | 18 | 1400 | |||
NM_021739.2 | NP_068507.2 | 20 | 1517 | |||
XM_006251170.2 | XP_006251232.1 | 23 | 1988 | |||
XM_006251171.2 | XP_006251233.1 | 22 | 1943 | |||
XM_006251172.2 | XP_006251234.1 | 22 | 1916 | |||
XM_006251174.2 | XP_006251236.1 | 21 | 1871 | |||
XM_006251176.2 | XP_006251238.1 | 19 | 1552 | |||
XM_006251177.2 | XP_006251239.1 | 19 | 1524 | |||
GenBank mRNA | CB692429.1 | - | 3 | 205 | ||
M16112.1 | - | 20 | 1497 | |||
X83375.1 | - | 20 | 1620 |
