siRNA Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042354.1 | NP_001035813.1 | 11 | 955 | ||
| NM_001042356.1 | NP_001035815.1 | 11 | 955 | |||
| NM_021739.2 | NP_068507.2 | 11 | 955 | |||
| XM_006251170.2 | XP_006251232.1 | 11 | 1054 | |||
| XM_006251171.2 | XP_006251233.1 | 11 | 1054 | |||
| XM_006251172.2 | XP_006251234.1 | 11 | 1054 | |||
| XM_006251174.2 | XP_006251236.1 | 11 | 1054 | |||
| XM_006251176.2 | XP_006251238.1 | 11 | 1035 | |||
| XM_006251177.2 | XP_006251239.1 | 11 | 1034 | |||
| GenBank mRNA | M16112.1 | - | 11 | 935 | ||
| X83375.1 | - | 11 | 917 | |||

Camk2b
Genomic Map