siRNA Details
| Gene Symbol: Camk2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012519.2 | NP_036651.1 | 18 | 1489 | ||
| XM_006233284.2 | XP_006233346.1 | 17 | 1648 | |||
| XM_006233285.2 | XP_006233347.1 | 17 | 1639 | |||
| XM_008761452.1 | XP_008759674.1 | 18 | 1708 | |||
| GenBank mRNA | BC107562.1 | - | 1249 | 7939424 | ||
| J05072.1 | - | 1489 | ||||
| L13406.1 | - | 4 | 226 | |||
| L13407.1 | - | 5 | 259 | |||
| L13408.1 | - | 5 | 286 | |||

Camk2d
Genomic Map