siRNA Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012520.2 | NP_036652.1 | 2 | 246 | ||
| GenBank mRNA | BC081853.1 | - | 2 | 143 | 7109889 | |
| FM043772.1 | - | 2 | 246 | |||
| M11670.1 | - | 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cat| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012520.2 | NP_036652.1 | 2 | 246 | ||
| GenBank mRNA | BC081853.1 | - | 2 | 143 | 7109889 | |
| FM043772.1 | - | 2 | 246 | |||
| M11670.1 | - | 2 | 197 | |||