siRNA Details
| Gene Symbol: Cel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016997.2 | NP_058693.2 | 10 | 1357 | ||
| GenBank mRNA | M15893.1 | - | 10 | 1357 | ||
| X16054.1 | - | 10 | 1353 | |||
| Z22803.1 | - | 9 | 1276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cel| Gene Symbol: Cel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016997.2 | NP_058693.2 | 10 | 1357 | ||
| GenBank mRNA | M15893.1 | - | 10 | 1357 | ||
| X16054.1 | - | 10 | 1353 | |||
| Z22803.1 | - | 9 | 1276 | |||