siRNA Details
| Gene Symbol: Cftr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031506.1 | NP_113694.1 | 17 | 2949 | ||
| XM_006236097.2 | XP_006236159.1 | 18 | 3068 | |||
| XM_008762680.1 | XP_008760902.1 | 18 | 3068 | |||
| GenBank mRNA | FJ959076.1 | - | 17 | 2707 | ||
| M89906.1 | - | 7 | 1369 | |||

Cftr
Genomic Map