siRNA Details
Gene Symbol: Chrm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012527.2 | NP_036659.2 | 5 | 671 | ||
XM_008771690.1 | XP_008769912.1 | 6 | 617 | |||
GenBank mRNA | AB017656.1 | - | 1 | 124 | ||
M16407.1 | - | 2 | 164 | |||
M18088.1 | - | 3 | 255 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
