siRNA Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270961.1 | NP_001257890.1 | 15 | 2633 | ||
| NM_012532.2 | NP_036664.2 | 15 | 2633 | |||
| XM_008760859.1 | XP_008759081.1 | 15 | 2821 | |||
| GenBank mRNA | AF202115.1 | - | 15 | 2600 | ||
| FQ220241.1 | - | 2 | 150 | |||
| L33869.1 | - | 15 | 2615 | |||

Cp
Genomic Map