siRNA Details
| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016998.3 | NP_058694.2 | 4 | 754 | ||
| GenBank mRNA | FQ232505.1 | - | 4 | 462 | ||
| J00713.1 | - | 4 | 754 | |||
| V01232.1 | - | 4 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa1| Gene Symbol: Cpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016998.3 | NP_058694.2 | 4 | 754 | ||
| GenBank mRNA | FQ232505.1 | - | 4 | 462 | ||
| J00713.1 | - | 4 | 754 | |||
| V01232.1 | - | 4 | 457 | |||