siRNA Details
| Gene Symbol: Cryaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289737.1 | NP_001276666.1 | 1 | 105 | ||
| NM_012534.4 | NP_036666.2 | 1 | 150 | |||
| XM_008772796.1 | XP_008771018.1 | 1 | 557 | |||
| GenBank mRNA | BC126082.1 | - | 1 | 150 | 8364912 | |
| U47922.1 | - | 1 | 103 | |||

Cryaa
Genomic Map