siRNA Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138874.2 | NP_620229.2 | 27 | 821 | ||
| GenBank mRNA | J00710.1 | - | 27 | 821 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csn1s1| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138874.2 | NP_620229.2 | 27 | 821 | ||
| GenBank mRNA | J00710.1 | - | 27 | 821 | ||