siRNA Details
| Gene Symbol: Ctrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012536.1 | NP_036668.1 | 5 | 340 | ||
| GenBank mRNA | FQ232114.1 | - | 5 | 362 | ||
| FQ233595.1 | - | 5 | 362 | |||
| FQ234337.1 | - | 5 | 362 | |||
| FQ234339.1 | - | 5 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctrb1| Gene Symbol: Ctrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012536.1 | NP_036668.1 | 5 | 340 | ||
| GenBank mRNA | FQ232114.1 | - | 5 | 362 | ||
| FQ233595.1 | - | 5 | 362 | |||
| FQ234337.1 | - | 5 | 362 | |||
| FQ234339.1 | - | 5 | 362 | |||