siRNA Details
| Gene Symbol: Cyp21a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057101.2 | NP_476442.2 | 2 | 233 | ||
| XM_006255907.2 | XP_006255969.1 | 2 | 310 | |||
| GenBank mRNA | U56853.1 | - | 2 | 234 | ||
| U56854.1 | - | 1 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp21a1| Gene Symbol: Cyp21a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057101.2 | NP_476442.2 | 2 | 233 | ||
| XM_006255907.2 | XP_006255969.1 | 2 | 310 | |||
| GenBank mRNA | U56853.1 | - | 2 | 234 | ||
| U56854.1 | - | 1 | 298 | |||