siRNA Details
| Gene Symbol: Cyp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044770.2 | NP_001038235.1 | 1 | 127 | ||
| GenBank mRNA | BC078684.1 | - | 1 | 127 | 7109305 | |
| BC098705.1 | - | 1 | 122 | 7106510 | ||
| Gene Symbol: Cyp4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175760.2 | NP_786936.1 | 1 | 123 | ||
| GenBank mRNA | M33936.1 | - | 1 | 123 | ||

Cyp4a2
Genomic Map