siRNA Details
| Gene Symbol: Dbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289982.1 | NP_001276911.1 | 3 | 740 | ||
| NM_001289983.1 | NP_001276912.1 | 2 | 629 | |||
| NM_012543.3 | NP_036675.1 | 3 | 1040 | |||
| GenBank mRNA | AY518348.1 | - | 3 | 339 | ||
| AY518349.1 | - | 2 | 228 | |||
| BC087668.1 | - | 3 | 1040 | 7317132 | ||
| FQ213620.1 | - | 3 | 554 | |||
| J03179.1 | - | 3 | 1006 | |||

Dbp
Genomic Map