siRNA Details
| Gene Symbol: Ddc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270852.1 | NP_001257781.1 | 10 | 1015 | ||
| NM_001270853.1 | NP_001257782.1 | 9 | 909 | |||
| NM_012545.4 | NP_036677.1 | 10 | 1055 | |||
| XM_006251474.2 | XP_006251536.1 | 10 | 1791 | |||
| XM_006251475.2 | XP_006251537.1 | 10 | 1631 | |||
| XM_008770251.1 | XP_008768473.1 | 10 | 1086 | |||
| XM_008770252.1 | XP_008768474.1 | 10 | 1149 | |||
| XM_008770253.1 | XP_008768475.1 | 9 | 966 | |||
| XM_008770254.1 | XP_008768476.1 | 10 | 1086 | |||
| GenBank mRNA | BC087032.1 | - | 10 | 1015 | 7098083 | |
| FQ219697.1 | - | 10 | 1053 | |||
| JF273828.1 | - | 9 | 909 | |||
| M27716.1 | - | 10 | 1060 | |||
| U31884.1 | - | 10 | 1037 | |||

Ddc
Genomic Map