siRNA Details
| Gene Symbol: Ednra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012550.2 | NP_036682.1 | 4, 5 | 1045 | ||
| XM_006255420.2 | XP_006255482.1 | 4, 5 | 1211 | |||
| XM_008772471.1 | XP_008770693.1 | 4, 5 | 918 | |||
| XM_008772472.1 | XP_008770694.1 | 3, 4 | 1113 | |||
| GenBank mRNA | DQ832324.1 | - | 3, 4 | 796 | ||
| DQ839232.1 | - | 2, 3 | 651 | |||
| DQ839234.1 | - | 4, 5 | 911 | |||
| M60786.1 | - | 3, 4 | 796 | |||

Ednra
Genomic Map