siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 10 | 1327 | ||
| XM_008770416.1 | XP_008768638.1 | 10 | 1406 | |||
| XM_008770417.1 | XP_008768639.1 | 10 | 1406 | |||
| XM_008770418.1 | XP_008768640.1 | 10 | 1405 | |||
| GenBank mRNA | HM801041.1 | - | 10 | 1292 | ||
| HM801042.1 | - | 10 | 1292 | |||
| M37394.2 | - | 10 | 1327 | |||

Egfr
Genomic Map