siRNA Details
| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 17 | 2178 | ||
| GenBank mRNA | M37394.2 | - | 17 | 2178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egfr| Gene Symbol: Egfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031507.1 | NP_113695.1 | 17 | 2178 | ||
| GenBank mRNA | M37394.2 | - | 17 | 2178 | ||