siRNA Details
| Gene Symbol: Cela1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012552.3 | NP_036684.1 | 5 | 458 | ||
| GenBank mRNA | FQ227752.1 | - | 5 | 480 | ||
| V01234.1 | - | 5 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cela1| Gene Symbol: Cela1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012552.3 | NP_036684.1 | 5 | 458 | ||
| GenBank mRNA | FQ227752.1 | - | 5 | 480 | ||
| V01234.1 | - | 5 | 479 | |||