siRNA Details
| Gene Symbol: Eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109908.1 | NP_001103378.1 | 4, 5 | 414 | ||
| NM_012554.3 | NP_036686.2 | 4, 5 | 365 | |||
| GenBank mRNA | BC063174.1 | - | 4, 5 | 358 | ||
| BC078896.1 | - | 4, 5 | 414 | 7112523 | ||
| BC081847.1 | - | 4, 5 | 338 | 7189453 | ||
| BC090069.1 | - | 4, 5 | 295 | 7108936 | ||
| BC091572.1 | - | 4, 5 | 335 | 7107492 | ||
| BC097343.1 | - | 4, 5 | 331 | 7461555 | ||
| CB809779.1 | - | 365 | ||||
| FQ225935.1 | - | 4, 5 | 378 | |||
| X02610.1 | - | 4, 5 | 354 | |||

Eno1
Genomic Map