siRNA Details
| Gene Symbol: Eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109908.1 | NP_001103378.1 | 7 | 807 | ||
| NM_012554.3 | NP_036686.2 | 7 | 758 | |||
| XM_006239444.1 | XP_006239506.1 | 7 | 774 | |||
| GenBank mRNA | BC063174.1 | - | 7 | 751 | ||
| BC078896.1 | - | 7 | 807 | 7112523 | ||
| BC081847.1 | - | 7 | 731 | 7189453 | ||
| BC090069.1 | - | 7 | 688 | 7108936 | ||
| BC091572.1 | - | 7 | 728 | 7107492 | ||
| BC097343.1 | - | 7 | 724 | 7461555 | ||
| FQ214707.1 | - | 7 | 680 | |||
| FQ225935.1 | - | 7 | 771 | |||
| X02610.1 | - | 7 | 747 | |||

Eno1
Genomic Map