siRNA Details
| Gene Symbol: Epo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017001.1 | NP_058697.1 | 3 | 207 | ||
| XM_006249101.2 | XP_006249163.1 | 2 | 349 | |||
| GenBank mRNA | D10763.1 | - | 3 | 207 | ||
| L10608.1 | - | 2 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epo| Gene Symbol: Epo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017001.1 | NP_058697.1 | 3 | 207 | ||
| XM_006249101.2 | XP_006249163.1 | 2 | 349 | |||
| GenBank mRNA | D10763.1 | - | 3 | 207 | ||
| L10608.1 | - | 2 | 176 | |||