siRNA Details
| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017002.2 | NP_058698.2 | 7, 8 | 987 | ||
| GenBank mRNA | BC089810.1 | - | 7, 8 | 987 | 7375416 | |
| D13566.1 | - | 7, 8 | 905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epor| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017002.2 | NP_058698.2 | 7, 8 | 987 | ||
| GenBank mRNA | BC089810.1 | - | 7, 8 | 987 | 7375416 | |
| D13566.1 | - | 7, 8 | 905 | |||