siRNA Details
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017003.2 | NP_058699.2 | 17 | 2080 | ||
| GenBank mRNA | AY116182.1 | - | 17 | 2080 | ||
| BC061863.1 | - | 17 | 2152 | 5597835 | ||
| X03362.1 | - | 17 | 1994 | |||
| Gene Symbol: LOC102552659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247245.2 | XP_006247307.2 | 4 | 343 | ||

Erbb2
Genomic Map