siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012557.2 | NP_036689.2 | 7, 8 | 931 | ||
| XM_006253488.2 | XP_006253550.1 | 8, 9 | 1206 | |||
| XM_006253489.2 | XP_006253551.1 | 7, 8 | 1063 | |||
| XM_006253490.2 | XP_006253552.1 | 7, 8 | 908 | |||
| XM_006253491.2 | XP_006253553.1 | 4, 5 | 1737 | |||
| XM_008771423.1 | XP_008769645.1 | 6, 7 | 2053 | |||
| GenBank mRNA | BC168147.1 | - | 7, 8 | 931 | 9024689 | |
| U73586.1 | - | 6, 7 | 707 | |||

Fancc
Genomic Map