siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012557.2 | NP_036689.2 | 13 | 1439 | ||
| XM_006253488.2 | XP_006253550.1 | 14 | 1714 | |||
| XM_006253489.2 | XP_006253551.1 | 13 | 1571 | |||
| XM_006253490.2 | XP_006253552.1 | 13 | 1416 | |||
| XM_006253491.2 | XP_006253553.1 | 10 | 2245 | |||
| XM_008771423.1 | XP_008769645.1 | 12 | 2561 | |||
| GenBank mRNA | BC168147.1 | - | 13 | 1439 | 9024689 | |
| U73586.1 | - | 12 | 1215 | |||

Fancc
Genomic Map