siRNA Details
| Gene Symbol: Fgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020071.2 | NP_064456.2 | 5, 6 | 846 | ||
| XM_006232524.2 | XP_006232586.1 | 5, 6 | 864 | |||
| XM_008761078.1 | XP_008759300.1 | 5, 6 | 803 | |||
| GenBank mRNA | AY321323.1 | - | 842 | |||
| AY325147.1 | - | 842 | ||||
| AY325153.1 | - | 842 | ||||
| BC088102.1 | - | 5, 6 | 846 | 7371665 | ||
| M27220.1 | - | 2, 3 | 258 | |||
| U05675.1 | - | 5, 6 | 847 | |||

Fgb
Genomic Map