siRNA Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012559.2 | NP_036691.2 | 7 | 730 | ||
| XM_006232525.1 | XP_006232587.1 | 7 | 884 | |||
| GenBank mRNA | BC078893.1 | - | 7 | 730 | 7122366 | |
| FQ209468.1 | - | 3 | 203 | |||
| FQ218666.1 | - | 5 | 532 | |||
| FQ219511.1 | - | 7 | 752 | |||
| FQ219541.1 | - | 7 | 751 | |||
| FQ219604.1 | - | 7 | 751 | |||
| FQ229132.1 | - | 7 | 757 | |||
| J00734.1 | - | 7 | 715 | |||

Fgg
Genomic Map