siRNA Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012559.2 | NP_036691.2 | 8 | 1144 | ||
| XM_006232525.1 | XP_006232587.1 | 8 | 1298 | |||
| GenBank mRNA | BC078893.1 | - | 8 | 1144 | 7122366 | |
| FQ209468.1 | - | 4 | 617 | |||
| FQ210959.1 | - | 2 | 402 | |||
| FQ218666.1 | - | 6 | 946 | |||
| FQ219408.1 | - | 8 | 1169 | |||
| FQ219511.1 | - | 8 | 1166 | |||
| FQ219541.1 | - | 8 | 1165 | |||
| FQ219604.1 | - | 8 | 1168 | |||
| FQ229132.1 | - | 8 | 1171 | |||
| J00734.1 | - | 8 | 1129 | |||

Fgg
Genomic Map