siRNA Details
| Gene Symbol: Gc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012564.3 | NP_036696.2 | 8 | 946 | ||
| GenBank mRNA | BC078891.1 | - | 8 | 946 | 7130137 | |
| FQ209652.1 | - | 8 | 957 | |||
| FQ210204.1 | - | 8 | 658 | |||
| FQ210565.1 | - | 1 | 40 | |||
| FQ211064.1 | - | 4 | 437 | |||
| FQ219041.1 | - | 5 | 567 | |||
| FQ219132.1 | - | 4 | 387 | |||
| J05148.1 | - | 8 | 922 | |||
| M12450.1 | - | 8 | 902 | |||

Gc
Genomic Map