siRNA Details
| Gene Symbol: Gc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012564.3 | NP_036696.2 | 8, 9 | 1090 | ||
| GenBank mRNA | BC078891.1 | - | 8, 9 | 1090 | 7130137 | |
| FQ209652.1 | - | 8, 9 | 1101 | |||
| FQ209886.1 | - | 8, 9 | 1100 | |||
| FQ210565.1 | - | 1, 2 | 184 | |||
| FQ210734.1 | - | 8, 9 | 1098 | |||
| FQ211064.1 | - | 4, 5 | 581 | |||
| FQ219041.1 | - | 5, 6 | 711 | |||
| FQ219081.1 | - | 8, 9 | 1101 | |||
| FQ219132.1 | - | 4, 5 | 531 | |||
| FQ219502.1 | - | 8, 9 | 1098 | |||
| J05148.1 | - | 8, 9 | 1066 | |||
| M12450.1 | - | 8, 9 | 1046 | |||

Gc
Genomic Map