siRNA Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017009.2 | NP_058705.2 | 7 | 1136 | ||
| XM_008768261.1 | XP_008766483.1 | 7 | 1202 | |||
| GenBank mRNA | BC088851.1 | - | 7 | 1136 | 7315338 | |
| EF094477.1 | - | 3 | 354 | |||
| L27219.1 | - | 7 | 1140 | |||
| U03700.1 | - | 7 | 1160 | |||

Gfap
Genomic Map